Learn More
1-(4-Hydroxyphenyl)ethanone Oxime (4-Hydroxyacetophenone Oxime), Mikromol™
Product Code. 30051328
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051328 25 mg 25mg
30049904 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051328 Supplier Mikromol Supplier No. MM0042.120025
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(4-Hydroxyphenyl)ethanone Oxime (4-Hydroxyacetophenone Oxime)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(4-Hydroxyphenyl)ethanone Oxime (4-Hydroxyacetophenone Oxime)
CAS 34523-34-7
Analyte or Component Names 1-(4-Hydroxyphenyl)ethanone Oxime (4-Hydroxyacetophenone Oxime)
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O2
Certifications/Compliance ISO 17025
Synonym 1-(4-Hydroxyphenyl)ethanone Oxime, Paracetamol Imp. G (EP), 4-Hydroxyacetophenone Oxime, Paracetamol Impurity G
InChI Formula InChI=1S/C8H9NO2/c1-6(9-11)7-2-4-8(10)5-3-7/h2-5,10-11H,1H3/b9-6-
SMILES C\C(=N\O)\c1ccc(O)cc1
IUPAC Name 4-[(Z)-N-hydroxy-C-methylcarbonimidoyl]phenol
Molecular Weight (g/mol) 151.16
Quantity 25 mg
Formula Weight 151.0633 g/mol