Learn More
1-(((5-(4-Nitrophenyl)furan-2-yl)methylene)amino)imidazolidine-2,4-dione, sodium salt hydrate(2:2:7), 98%, BLD Pharm™
Product Code. 30948621
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
25 g
Unit Size:
100mg
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948621 1 g 1g
30931309 25 g 25g
30945671 5 g 5g
30931699 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30948621 Supplier BLD Pharm Supplier No. BD1234381g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 24868-20-0
Molecular Formula C28H32N8Na2O17
MDL Number MFCD07357270
Quantity 1 g
SMILES [H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].O=C(CN1N=CC2=CC=C(O2)C3=CC=C(C=C3)[N+]([O-])=O)[N-]C1=O.O=C(CN4N=CC5=CC=C(O5)C6=CC=C(C=C6)[N+]([O-])=O)[N-]C4=O.[Na+].[Na+]
Molecular Weight (g/mol) 798.58
Percent Purity 98%
Chemical Name or Material 1-(((5-(4-Nitrophenyl)furan-2-yl)methylene)amino)imidazolidine-2,4-dione, sodium salt hydrate(2:2:7)