Learn More
1,5-Dichloroanthraquinone, TRC
Product Code. 30378808
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378808 1 g -
30381751 10 g -
30376643 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378808 Supplier Toronto Research Chemicals Supplier No. TRCD4319951G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 82-46-2
Molecular Formula C14H6Cl2O2
Molecular Weight (g/mol) 277.1
Synonym 1,5-Dichloro-9,10-anthracenedione, 1,5-Dichloro-9,10-anthraquinone, 1,5-Dichloroanthraquinone, NSC 13969
SMILES Clc1cccc2C(=O)c3c(Cl)cccc3C(=O)c12
Specifications
Quantity 1 g
InChI Formula InChI=1S/C14H6Cl2O2/c15-9-5-1-3-7-11(9)14(18)8-4-2-6-10(16)12(8)13(7)17/h1-6H
Chemical Name or Material 1,5-Dichloroanthraquinone