Learn More
1-(5-Methylfuran-2-yl)propan-1-one, TRC
Product Code. 30482802
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482802 500 mg 500mg
30474540 50 mg 50mg
30475283 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 30482802 Fournisseur Toronto Research Chemicals Code fournisseur TRCM305803500MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10599-69-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.16
Synonym 1-Propanone, 1-(5-methyl-2-furyl)- (6CI,7CI,8CI), 1-(5-Methyl-2-furanyl)-1-propanone1-(5-Methyl-2-furyl)propan-1-one, 2-Methyl-5-propionylfuran, 5-Methyl-2-propanoylfuran, 5-Methyl-2-propionylfuran
IUPAC Name 1-(5-methylfuran-2-yl)propan-1-one
SMILES CCC(=O)c1oc(C)cc1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C8H10O2/c1-3-7(9)8-5-4-6(2)10-8/h4-5H,3H2,1-2H3
Formula Weight 138.0681
Chemical Name or Material 1-(5-Methylfuran-2-yl)propan-1-one