Learn More
1,6-Diphenyl-1,3,5-hexatriene, TRC
Product Code. 30135391
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135391 1 g 1g
30004944 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135391 Supplier Toronto Research Chemicals Supplier No. TRCD4898781G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1720-32-7
Molecular Formula C18 H16
Molecular Weight (g/mol) 232.32
Synonym 1,6-Diphenylhexatriene, DPH, DPH (dye), Diphenylhexatriene, NSC 90479, alpha,ω-Diphenylhexatriene, 1,1'-(1,3,5-Hexatriene-1,6-diyl)bis[benzene];
IUPAC Name 6-phenylhexa-1,3,5-trienylbenzene
SMILES C(=CC=Cc1ccccc1)C=Cc2ccccc2
Specifications
Quantity 1 g
InChI Formula InChI=1S/C18H16/c1(5-11-17-13-7-3-8-14-17)2-6-12-18-15-9-4-10-16-18/h1-16H
Chemical Name or Material 1,6-Diphenyl-1,3,5-hexatriene