Learn More
1,8-Dihydroxy-3-(hydroxymethyl)-anthracene-9,10-dione (Aloe-Emodin), Mikromol™
Product Code. 17912996
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17912996 100 mg 100mg
17931176 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17912996 Supplier Mikromol Supplier No. MM1318.02
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,8-Dihydroxy-3-(hydroxymethyl)-anthracene-9,10-dione (Aloe-Emodin)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Aloe Emodin
CAS 481-72-1
Analyte or Component Names Aloe Emodin
Shipping Condition Room Temperature
Molecular Formula C15 H10 O5
Certifications/Compliance ISO 17025
Synonym Diacerein Imp. B (EP), 1,8-Dihydroxy-3-(hydroxymethyl)-9,10-anthracenedione, 1,8-Dihydroxy-3-hydroxymethylanthraquinone, 3-Hydroxymethylchrysazin, 3-Hydroxymethylchrysazine, Aloe emodin, Aloe-emodol, NSC 38628, Rhabarberone, Diacerein Impurity B
InChI Formula InChI=1S/C15H10O5/c16-6-7-4-9-13(11(18)5-7)15(20)12-8(14(9)19)2-1-3-10(12)17/h1-5,16-18H,6H2
SMILES OCc1cc(O)c2C(=O)c3c(O)cccc3C(=O)c2c1
IUPAC Name 1,8-dihydroxy-3-(hydroxymethyl)anthracene-9,10-dione
Molecular Weight (g/mol) 270.24
Quantity 100 mg
Formula Weight 270.0528 g/mol