Learn More
1-Benzofuran-3-ylacetonitrile, 98%, BLD Pharm™
Product Code. 31007266
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31007266 1 g 1g
30998778 250 mg 250mg
30992823 5 g 5g
31005566 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31007266 Supplier BLD Pharm Supplier No. BD126121g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 52407-43-9
Molecular Formula C10H7NO
MDL Number MFCD02183555
Quantity 1 g
SMILES N#CCC1=COC2=C1C=CC=C2
Molecular Weight (g/mol) 157.17
Percent Purity 98%
Chemical Name or Material 1-Benzofuran-3-ylacetonitrile