missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chlorobenzene, 98+%

Product Code. 11449353
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11449353

Brand: Thermo Scientific Alfa Aesar A11782.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 694-80-4 | C6H4BrCl | 191.452 g/mol

1-Bromo-2-chlorobenzene finds application in Gas Chromatography and Liquid Chromatography analysis. It is also used as an intermediate in pharmaceutical and synthetic chemistry. Further, it acts as a reagent in the preparation of arylpiperidines and aryltetrahydropyridines as 5-HT2C agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-chlorobenzene finds application in Gas Chromatography and Liquid Chromatography analysis. It is also used as an intermediate in pharmaceutical and synthetic chemistry. Further, it acts as a reagent in the preparation of arylpiperidines and aryltetrahydropyridines as 5-HT2C agonists.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-80-4
Molecular Formula C6H4BrCl
Molecular Weight (g/mol) 191.452
MDL Number MFCD00000532
InChI Key QBELEDRHMPMKHP-UHFFFAOYSA-N
Synonym 2-bromochlorobenzene, o-bromochlorobenzene, 2-chlorobromobenzene, o-chlorobromobenzene, benzene, 1-bromo-2-chloro, 2-bromo-1-chlorobenzene, 1-chloro-2-bromobenzene, benzene, bromochloro, unii-pec7z3yx6p, 1-bromo-2-chloro-benzene
PubChem CID 12754
IUPAC Name 1-bromo-2-chlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Br

Specifications

Melting Point -13°C
Density 1.649
Boiling Point 203°C to 205°C
Quantity 1000 g
Refractive Index 1.5815
Beilstein 1855303
Solubility Information Immiscible with water.
Formula Weight 191.46
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-2-chlorobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.