Learn More
1-Bromodocosane, TRC
Spécification
Spécification
| Chemical Name or Material | 1-Bromodocosane |
| CAS | 6938-66-5 |
| Molecular Formula | C22H45Br |
| Quantity | 250 mg |
| Synonym | Behenyl Bromide, Docosyl Bromide, NSC 53991, S 71281 |
| InChI Formula | InChI=1S/C22H45Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23/h2-22H2,1H3 |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCBr |
| IUPAC Name | 1-bromodocosane |
| Molecular Weight (g/mol) | 389.5 |
| Formula Weight | 388.2705 |