Learn More
1-Furfurylpyrrole, 99%, Thermo Scientific™
Product Code. 11436316
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436316 100 g 100g
11416316 5 g 5g
11426316 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436316 Supplier Thermo Scientific Alfa Aesar Supplier No. B23105.22
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 1438-94-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00010080
InChI Key BTBFUBUCCJKJOZ-UHFFFAOYSA-N
Synonym 1-furfurylpyrrole, n-furfurylpyrrole, 1-furan-2-ylmethyl-1h-pyrrole, 1-furfuryl-1h-pyrrole, 1h-pyrrole, 1-2-furanylmethyl, 1-2-furanylmethyl-1h-pyrrole, n-2-furfuryl pyrrole, n-furfuryl pyrrole, pyrrole, 1-furfuryl, 1-2-furfuryl pyrrole
PubChem CID 15037
IUPAC Name 1-(furan-2-ylmethyl)pyrrole
SMILES C(N1C=CC=C1)C1=CC=CO1
Spécification
Density 1.081
Boiling Point 76°C to 78°C (1 mmHg)
Flash Point 92°C (199°F)
Refractive Index 1.531
Quantity 100 g
UN Number UN2810
Sensitivity Light sensitive
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material 1-Furfurylpyrrole