Learn More
1-Heptanesulfonic acid sodium salt monohydrate, ∽98%, MP Biomedicals™
Product Code. 17861371 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17861371 25 g 25g
17861361 5 g 5g
11453791 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17861371 Supplier MP Biomedicals Supplier No. 0215123925
  • ONLINE EXCLUSIVE PRICE
    302.00€
    240.45€ / 25g
    Économisez 61.55€
    20% de réduction
 Restricted Item
Expédition estimée: 30-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Item Name 1-Heptanesulfonic Acid, Sodium Salt, Monohydrate
Form Crystalline Powder
Quantity 25 g
Appearance White