Learn More
1-Iodo-3-(trifluoromethyl)benzene, 98%, BLD Pharm™
Product Code. 30985499
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30985499 100 g
30971876 10 g
30984027 500 g
30974378 1 kg
30970764 5 g
30976194 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30985499 Supplier BLD Pharm Supplier No. BD10288100g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
(Stabilized with Copper chip)
Specifications
CAS 401-81-0
Molecular Formula C7H4F3I
MDL Number MFCD00001049
Quantity 100 g
SMILES FC(C1=CC(I)=CC=C1)(F)F
Molecular Weight (g/mol) 272.01
Percent Purity 98%
Chemical Name or Material 1-Iodo-3-(trifluoromethyl)benzene