Learn More
1-(Mercaptomethyl)cyclopropaneacetic Acid, TRC
Product Code. 30465297
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30465297 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465297 Supplier Toronto Research Chemicals Supplier No. TRCM2535701G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(Mercaptomethyl)cyclopropaneacetic Acid
CAS 162515-68-6
Molecular Formula C6 H10 O2 S
Quantity 1 g
Synonym 1-(Mercaptomethyl)cyclopropaneacetic Acid, [1-(Mercaptomethyl)cyclopropyl]acetic Acid
InChI Formula InChI=1S/C6H10O2S/c7-5(8)3-6(4-9)1-2-6/h9H,1-4H2,(H,7,8)
SMILES OC(=O)CC1(CS)CC1
IUPAC Name 2-[1-(sulfanylmethyl)cyclopropyl]acetic acid
Molecular Weight (g/mol) 146.21
Formula Weight 146.0402