Learn More
1-Methyl Mebendazole, TRC
Product Code. 30477405
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477405 10 mg 10mg
30466650 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477405 Supplier Toronto Research Chemicals Supplier No. TRCM32015010MG
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 132119-11-0
Molecular Formula C17 H15 N3 O3
Molecular Weight (g/mol) 309.32
Synonym Methyl (5-Benzoyl-1-methyl-1H-benzimidazol-2-yl)carbamate, Mebendazole Imp. D (EP)
IUPAC Name methyl N-(5-benzoyl-1-methylbenzimidazol-2-yl)carbamate
SMILES COC(=O)Nc1nc2cc(ccc2n1C)C(=O)c3ccccc3
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C17H15N3O3/c1-20-14-9-8-12(15(21)11-6-4-3-5-7-11)10-13(14)18-16(20)19-17(22)23-2/h3-10H,1-2H3,(H,18,19,22)
Formula Weight 309.1113
Percent Purity >95%
Chemical Name or Material Methyl (5-Benzoyl-1-methyl-1H-benzimidazol-2-yl)carbamate
Grade HPLC