missing translation for 'onlineSavingsMsg'
Learn More

1-Methylisatin, 97%

Product Code. 11489773
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11489773

Brand: Thermo Scientific Alfa Aesar A12942.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2058-74-4 | C9H7NO2 | 161.16 g/mol

1-Methylisatin is used as a reactant for stereoselective preparation of spirobicyclic and bis-spirotricyclic pyrazolidinones, for regioselective preparation of spirocyclic oxindole-butenolides, for synthesis of spiro-oxindoles, for preparation of unsymmetrical oxindoles, for stereoselective preparation of hydroxyloxindoles via Morita-Baylis-Hillman reaction, for preparation of pyridinecarboxylic acid [(oxo)indolylidene)hydrazide derivatives (Schiff base hydrazides) as antibacterial agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylisatin is used as a reactant for stereoselective preparation of spirobicyclic and bis-spirotricyclic pyrazolidinones, for regioselective preparation of spirocyclic oxindole-butenolides, for synthesis of spiro-oxindoles, for preparation of unsymmetrical oxindoles, for stereoselective preparation of hydroxyloxindoles via Morita-Baylis-Hillman reaction, for preparation of pyridinecarboxylic acid [(oxo)indolylidene)hydrazide derivatives (Schiff base hydrazides) as antibacterial agents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2058-74-4
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005812
InChI Key VCYBVWFTGAZHGH-UHFFFAOYSA-N
Synonym n-methylisatin, 1-methylisatin, 1-methyl-1h-indole-2,3-dione, 1-methylindoline-2,3-dione, 1h-indole-2,3-dione, 1-methyl, 1-methyl-indole-2,3-dione, 1 methylisatine, n-methylindol-2,3-dione, 1-methyl-2,3-dihydro-1h-indole-2,3-dione, 1-methyl-2,3-indolinedione
PubChem CID 16358
SMILES CN1C(=O)C(=O)C2=CC=CC=C12

Specifications

Melting Point 129°C to 133°C
Quantity 5 g
Beilstein 128280
Solubility Information Insoluble in water.
IUPAC Name 1-methyl-2,3-dihydro-1H-indole-2,3-dione
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 1-Methylisatin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.