Learn More
1-Nitro-3,5-bis(trifluoromethyl)benzene, 98%, BLD Pharm™
Product Code. 30960257
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30960257 5 g 5g
30960256 25 g -
30960056 100 g 100g
30951279 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30960257 Supplier BLD Pharm Supplier No. BD108035g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 328-75-6
Molecular Formula C8H3F6NO2
MDL Number MFCD00000384
Quantity 5 g
SMILES FC(C1=CC(C(F)(F)F)=CC([N+]([O-])=O)=C1)(F)F
Molecular Weight (g/mol) 259.11
Percent Purity 98%
Chemical Name or Material 1-Nitro-3,5-bis(trifluoromethyl)benzene