Learn More
1-Pyrenebutyric acid, 95%, BLD Pharm™
Product Code. 30968356
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30968356 5 g 5g
30950641 10 g 10g
30950158 25 g 25g
30961092 100 g 100g
30965061 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30968356 Supplier BLD Pharm Supplier No. BD11565g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 3443-45-6
Molecular Formula C20H16O2
MDL Number MFCD00004141
Quantity 5 g
SMILES O=C(O)CCCC1=C(C2=C34)C=CC4=CC=CC3=CC=C2C=C1
Molecular Weight (g/mol) 288.34
Percent Purity 95%
Chemical Name or Material 1-Pyrenebutyric acid