Learn More
11-Bromoundecanoic Acid, TRC
Product Code. 30344502
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30344502 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344502 Supplier Toronto Research Chemicals Supplier No. TRCB6888805G
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 11-Bromoundecanoic Acid
CAS 2834-05-1
Molecular Formula C11H21BrO2
Quantity 5 g
Synonym NSC 14781, ω-Bromoundecanoic Acid
InChI Formula InChI=1S/C11H21BrO2/c12-10-8-6-4-2-1-3-5-7-9-11(13)14/h1-10H2,(H,13,14)
SMILES OC(=O)CCCCCCCCCCBr
IUPAC Name 11-bromoundecanoic acid
Molecular Weight (g/mol) 265.19
Formula Weight 264.0725