Learn More
Draeger Safety 112 L Test Gases
Product Code. 15950931
Change view
Click to view available options
Capacity (Metric):
112 L
Unit Size:
1.60 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15950931 112 L 1.60 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15950931 Supplier Draeger Safety Supplier No. 6812377
  • ONLINE EXCLUSIVE PRICE
    1006.43€
    523.19€ / 1.6L
    Économisez 483.24€
    48% de réduction
 Restricted Item
Expédition estimée: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Draeger Safety Calibration and test gas mixture consisting of 15 ppm H2S (hydrogen sulfide), 50 ppm CO (carbon monoxide), 2 vol.% CO2 (carbon dioxide), 2.5 vol.% CH4 (methane), 18 vol.% O2 ( Oxygen) in nitrogen in a disposable bottle made of aluminum.
  • Nominal volume: 1.6 L
  • Consisting of: 15 ppm H2S (hydrogen sulfide), 50 ppm CO (carbon monoxide), 2 vol.% CO2 (carbon dioxide), 2.5 vol.% CH4 (methane), 18 vol.% O2 ( Oxygen) in nitrogen in a disposable bottle made of aluminum
Specifications
Capacity (Metric) 112 L
Pressure Range 70 bar