Learn More
Thermo Scientific™ AI/AS 1610 Liquid Autosampler
Product Code. 17823204 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
AI 1610 Liquid Autosampler
AS 1610-155 Liquid Autosampler
Sample Capacity:
155 x 2 mL Vials
8 x 2 mL Vials
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Sample Capacity Unit Size
17823204 AS 1610-155 Liquid Autosampler 155 x 2 mL Vials Each
17881810 AI 1610 Liquid Autosampler 8 x 2 mL Vials Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17823204 Supplier Thermo Scientific™ Supplier No. MI2510000003
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
The Thermo Scientific™ AI/AS 1610 Autosampler elevates user experience on liquid sampling automation for GC and GC-MS through simplicity, robustness, and throughput capability.
Enhance liquid sampling automation for your gas chromatography (GC) applications with simplicity and robustness. Designed to meet the demands of any busy high-throughput lab, the Thermo Scientific™ AI/AS 1610 Liquid Autosampler will help to process more samples efficiently and safely with its advanced design and usability features, greatly enhanced for easy adoption for both beginners and experienced users. With the possibility to expand to up to a 310-sample capacity with simultaneous dual-tower operations, the AI/AS 1610 offers the fit-for-purpose solution to budget-conscious laboratories looking for reliable automation of liquid samples injection into a GC or GC-MS system.
Combine performance, simplicity, and robustness for your every-day workflow.
The AI/AS 1610 Liquid Autosampler is available in two models: the upgradable 8-vial capacity AI 1610 and the 155-vial capacity AS 1610. Both offers the dual-tower Gemini configuration for simultaneous injection in two channels.
Enhanced usability
  • Easy set up through self-aligning not requiring any position adjustment
  • Easy slide-in installation and free access to the injector for maintenance
  • Easy syringe viewing and replacement with illuminated compartment and magnifier lens
  • Enhanced color-coded instrument status indication
  • Safe operation with automatic selection of the best injection settings through recognition of the injector type and injection mode
  • Simplified connectivity with a single cable including communication, synchronization, and power supply
Enhanced sample throughput
  • Wide sample capacity scalability to fit any workload demand
  • Dual Tower Gemini for upmost productivity with simplified configuration and connectivity
Enhanced performance
  • Expanded applicability with liquid sample volume injection up to 80μL
  • Free pre- and post-injection syringe rinsing with single or combined solvents (up to four) virtually eliminates sample carry-over
  • Efficient, accurate sampling of low boiling compounds with the syringe kept away from the inlet’s temperature influence
  • Seamless integration into Thermo Scientific™ chromatography and™ software
Specifications
Sample Capacity 155 x 2 mL Vials
Injection Mode Automatically selected and optimized according to injector type
Injection Speed Fast/Slow/Variable (1 to 60,000.00 μL/min.)
Injection Volume 0.01 to 80 μL
Description AS 1610-155 Liquid Autosampler
For Use With (Equipment) TRACE 1600 Series
Washing Solvents Up to 4
Quantity Each