Learn More
17-(Allylamino)-17-demethoxygeldanamycin, 99%
Description
17-(Allylamino)-17-demethoxygeldanamycin (17-AAG) inhibits heat shock protein 90 (Hsp90), the expression of heat shock factor-1, and the activity of oncogenic proteins such as N-ras, Ki-ras, c-Akt, and p185erB2. In addition to its anti-tumor effects, 17-AAG also induces apoptosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in DMSO at 150 mg/mL; soluble in ethanol at 5 mg/mL. Very poorly soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Specifications
| Melting Point | 207°C to 209°C |
| Flash Point | >110°C (230°F) |
| Quantity | 200 mg |
| Sensitivity | Light sensitive |
| Solubility Information | Soluble in DMSO at 150mg/mL; soluble in ethanol at 5mg/mL; Very poorly soluble in water |
| Formula Weight | 585.69 |
| Percent Purity | 99% |
| Physical Form | Solid |
| Chemical Name or Material | 17-(Allylamino)-17-demethoxygeldanamycin |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.