Learn More
1H,1H,2H,2H-Perfluorooctyl-1-thiol, TRC
Product Code. 30415889
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415889 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415889 Supplier Toronto Research Chemicals Supplier No. TRCP2861255G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1H,1H,2H,2H-Perfluorooctyl-1-thiol
CAS 34451-26-8
Purity Grade Notes GC
Molecular Formula C8H5F13S
Quantity 5 g
Synonym 1-Octanethiol','3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro- (9CI','ACI): 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanethiol (ACI), 1,1,2,2-Tetrahydroperfluorooctanethiol, 1,1,2,2-Tetrahydroperfluorooctyl mercaptan, 1H,1H,1H,2H-Perfluorooctanethiol, 1H,1H,2H,2H-Perfluorooctanethiol, 2-(Perfluorohexyl)ethane-1-thiol, 2-(Perfluorohexyl)ethanethiol, Foralkyl EM 6
InChI Formula InChI=1S/C8H5F13S/c9-3(10,1-2-22)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h22H,1-2H2
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCS
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctane-1-thiol
Molecular Weight (g/mol) 380.17
Formula Weight 379.9904
Percent Purity >95