Learn More
1H-indazole-3-carboxylic acid, Thermo Scientific™
Product Code. 10545503
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10545503 1 g 1g
10802621 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10545503 Supplier Thermo Scientific Supplier No. MO07468DA
 Restricted Item
Expédition estimée: 02-12-2026
Add to basket
This item is not returnable. View return policy
Chemical Identifiers
CAS 4498-67-3
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
InChI Key BHXVYTQDWMQVBI-UHFFFAOYSA-N
Synonym indazole-3-carboxylic acid, 3-carboxyindazole, 3-indazolecarboxylic acid, indazole-3-carboxylicacid, 1h-indazol-3-carbonic acid, 2h-indazole-3-carboxylic acid, 3-carboxy-1h-indazole, 1h-indazole-3-carbonic acid, 1-h-indazole-3-carboxylic acid, benzopyrazole-3-carboxylic acid
PubChem CID 78250
IUPAC Name 1H-indazole-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=NN2)C(=O)O
Specifications
Assay Percent Range 97%
Quantity 1 g
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 1H-Indazole-3-carboxylic acid