Learn More
(1R,2R,3S,5R)-(-)-2,3-Pinanediol, 99%, Thermo Scientific™
Product Code. 15473078
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473078 1 g 1g
15483078 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473078 Supplier Thermo Scientific Alfa Aesar Supplier No. H56580.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22422-34-0
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD09955216
InChI Key MOILFCKRQFQVFS-UHFFFAOYNA-N
Synonym 1r,2r,3s,5r---2,3-pinanediol, 1r,2r,3s,5r---pinanediol, 1r,2r,3s,5r-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, --2,3-pinanediol, unii-r58l0w3a75, --cis-pinane-cis-2,3-diol, pinanediol, --2-hydroxyisopinocampheol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1r,2r,3s,5r
PubChem CID 6553875
IUPAC Name (1R,3S,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol
SMILES CC1(C)C2CC1C(C)(O)C(O)C2
Specifications
Melting Point 57°C to 59°C
Boiling Point 101°C to 102°C (1 mmHg)
Flash Point 110°C (230°F)
Quantity 1 g
Beilstein 2039144
Optical Rotation −8.6° (c=6.5 in Toluene)
Formula Weight 170.25
Percent Purity 99%
Chemical Name or Material (1R,2R,3S,5R)-(-)-2,3-Pinanediol