Learn More
Alfa Aesar™ (1R)-(-)-Camphorquinone, 98%
Product Code. 11461097
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461097 5 g 5g
11451097 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461097 Supplier Alfa Aesar™ Supplier No. B24092.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Chemical Identifiers
CAS 10334-26-6
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00064160,MFCD00082863,MFCD00064160,MFCD00082863
InChI Key VNQXSTWCDUXYEZ-LDWIPMOCSA-N
Synonym 1r,4s-1,7,7-trimethylnorbornane-2,3-dione, 1r,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, 1s,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione
PubChem CID 16211789
IUPAC Name (4R)-4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
SMILES CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2=O
Specifications
Melting Point 197°C to 202°C
Quantity 5 g
Beilstein 2327696
Optical Rotation −102° (c=2 in Ethanol)
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material (1R)-(-)-Camphorquinone
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.