Learn More
2,3',4'-Trichloroacetophenone, TRC
Spécification
Spécification
| Chemical Name or Material | 2,3',4'-Trichloroacetophenone |
| CAS | 42981-08-8 |
| Molecular Formula | C8H5OCl3 |
| Quantity | 50 mg |
| Synonym | 2-Chloro-1-(3,4-dichlorophenyl)ethanone, 2-Chloro-1-(3,4-dichlorophenyl)ethan-1-one, 2-Chloro-3',4'-dichloroacetophenone3,4-Dichlorophenacyl Chloride, NSC 243689’ |
| InChI Formula | InChI=1S/C8H5Cl3O/c9-4-8(12)5-1-2-6(10)7(11)3-5/h1-3H,4H2 |
| SMILES | ClCC(=O)c1ccc(Cl)c(Cl)c1 |
| IUPAC Name | 2-chloro-1-(3,4-dichlorophenyl)ethanone |
| Molecular Weight (g/mol) | 223.48 |
| Formula Weight | 221.9406 |