Learn More
2,3-Dihydrobenzo[b]furan-5-carbaldehyde, 95%, Thermo Scientific™
Product Code. 10512672
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10512672 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10512672 Supplier Thermo Scientific Maybridge Supplier No. CC00804DA
 Restricted Item
Expédition estimée: 16-12-2026
Add to basket
This item is not returnable. View return policy
Specifications
CAS 55745-70-5
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C9H8O2
MDL Number MFCD00068058
Quantity 1 g
Synonym 2,3-dihydrobenzofuran-5-carboxaldehyde, 2,3-dihydrobenzo b furan-5-carbaldehyde, 2,3-dihydrobenzofuran-5-carbaldehyde, 2,3-dihydrobenzo b furan-5-carboxaldehyde, 2,3-dihyro-1-benzofuran-5-carbaldehyde, 2,3-dihydro-benzofuran-5-carbaldehyde, 5-coumarancarboxaldehyde, 2,3-dihydro-5-benzofurancarboxaldehyde, 5-benzofurancarboxaldehyde, 2,3-dihydro, 2,3-dihydro-5-benzofurancarbaldehyde
InChI Key WEBVDBDZSOJGPB-UHFFFAOYSA-N
SMILES O=CC1=CC=C2OCCC2=C1
IUPAC Name 2,3-dihydro-1-benzofuran-5-carbaldehyde
Molecular Weight (g/mol) 148.16
PubChem CID 735901
Formula Weight 148.16
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-5-carbaldehyde