Learn More
2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, 98%, BLD Pharm™
Product Code. 31032559
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31032559 100 g
31026794 1 kg
31030947 500 g
31032401 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31032559 Supplier BLD Pharm Supplier No. BD126434100g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 10191-41-0
Molecular Formula C29H50O2
MDL Number MFCD00006848
Quantity 100 g
SMILES CC1=C2C(OC(CCCC(CCCC(CCCC(C)C)C)C)(C)CC2)=C(C(C)=C1O)C
Molecular Weight (g/mol) 430.71
Percent Purity 98%
Chemical Name or Material 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol