Learn More
2-[(5-Bromo-2-methylphenyl)methyl]-5-(4-fluorophenyl)thiophene, TRC
Product Code. 30348403
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348403 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348403 Supplier Toronto Research Chemicals Supplier No. TRCB685970250MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-[(5-Bromo-2-methylphenyl)methyl]-5-(4-fluorophenyl)thiophene
CAS 1030825-20-7
Molecular Formula C18H14BrFS
Quantity 250 mg
Synonym 2-(5-Bromo-2-methylbenzyl)-5-(4-fluorophenyl)thiophene
InChI Formula InChI=1S/C18H14BrFS/c1-12-2-5-15(19)10-14(12)11-17-8-9-18(21-17)13-3-6-16(20)7-4-13/h2-10H,11H2,1H3
SMILES Cc1ccc(Br)cc1Cc2ccc(s2)c3ccc(F)cc3
IUPAC Name 2-[(5-bromo-2-methylphenyl)methyl]-5-(4-fluorophenyl)thiophene
Molecular Weight (g/mol) 361.27
Formula Weight 359.9984