Learn More
2-(5-Chlorobenzo[b]thiophen-3-yl)acetic acid, 98%, BLD Pharm™
Product Code. 30803605
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30803605 250 mg 250mg
30800153 1 g 1g
30793563 100 mg 100mg
30792204 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30803605 Supplier BLD Pharm Supplier No. BD12581250mg
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 17266-30-7
Molecular Formula C10H7ClO2S
MDL Number MFCD00052308
Quantity 250 mg
SMILES O=C(O)CC1=CSC2=CC=C(Cl)C=C21
Molecular Weight (g/mol) 226.68
Percent Purity 98%
Chemical Name or Material 2-(5-Chlorobenzo[b]thiophen-3-yl)acetic acid