Learn More
2,5-dimethylphenyl isothiocyanate, Thermo Scientific™
Product Code. 10681365
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10681365 10 g 10g
10648755 5 g 5g
10337164 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10681365 Supplier Thermo Scientific Maybridge Supplier No. SB01618EA
 Restricted Item
Expédition estimée: 26-11-2026
Add to basket
This item is not returnable. View return policy
Chemical Identifiers
CAS 19241-15-7
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
InChI Key RCBXZRJVMSWGAO-UHFFFAOYSA-N
PubChem CID 140499
IUPAC Name 2-isothiocyanato-1,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C)N=C=S
Specifications
Quantity 10 g
Assay Percent Range 97%
Synonym 2,5-dimethylphenyl isothiocyanate, benzene,2-isothiocyanato-1,4-dimethyl, 2,5-dimethylbenzenisothiocyanate, 2-isothiocyanato-1,4-dimethyl-benzene, 2,5-xylyl isothiocyanate, 2,5-dimethylphenyl-isothiocyanate, 2,5-dimethylphenylisothiocyanate, #, benzene, 2-isothiocyanato-1,4-dimethyl
Formula Weight 163.24
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethylphenyl isothiocyanate