Learn More
2,6-Dimethylphenylisocyanate, 95%, BLD Pharm™
Product Code. 30966443
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30966443 1 g 1g
30949653 5 g 5g
30959431 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30966443 Supplier BLD Pharm Supplier No. BD116951g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 28556-81-2
Molecular Formula C9H9NO
MDL Number MFCD00002012
Quantity 1 g
SMILES CC1=C(N=C=O)C(C)=CC=C1
Molecular Weight (g/mol) 147.17
Percent Purity 95%
Chemical Name or Material 2,6-Dimethylphenylisocyanate