Learn More
MP Biomedicals™ 2-8 CELLsium™ Medium for Short-Term Cell Storage
Product Code. 16399855 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16399855 100 mL 100mL
16309865 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16399855 Supplier MP Biomedicals™ Supplier No. 092780349
  • ONLINE EXCLUSIVE PRICE
    308.98€
    268.80€ / 100mL
    Économisez 40.18€
    13% de réduction
 Restricted Item
Expédition estimée: 06-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
2-8 CELLsius is a cytoprotective, protein free, ready to use bio-solution without DMSO for the short-term storage of cells or tissue.
  • For short-term storage, add cold (2 to 8 °C) 2-8 CELLsium at a recommended cell concentration of 1 x 106 mL to 8 x 106 mL
  • The 2-8 CELLsium can be removed after centrifugation, cell concentration adjusted and transferred to an appropriate culture flask with growth medium
  • It will keep the cells healthy while in a temporary senescent phase, unlike regular growth media, which cannot
Specifications
Form Liquid
Sterility Sterile
For Use With (Application) Short-term cell storage
Quantity 100 mL
Content And Storage Stored at 2 to 8°C