Learn More
2-Amino-1-naphthalenesulfonic Acid, TRC
Product Code. 30134956
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134956 250 g 250g
30132538 100 g 100g
17938334 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134956 Supplier Toronto Research Chemicals Supplier No. TRCA618330250G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 81-16-3
Molecular Formula C10 H9 N O3 S
Molecular Weight (g/mol) 223.25
Synonym Tobias Acid, 2-Naphthylamine-1-sulfonic Acid, 1-Sulfo-2-naphthylamine
IUPAC Name 2-aminonaphthalene-1-sulfonic acid
SMILES Nc1ccc2ccccc2c1S(=O)(=O)O
Specifications
Quantity 250 g
InChI Formula InChI=1S/C10H9NO3S/c11-9-6-5-7-3-1-2-4-8(7)10(9)15(12,13)14/h1-6H,11H2,(H,12,13,14)
Formula Weight 223.0303
Percent Purity >95%
Chemical Name or Material 2-Amino-1-naphthalenesulfonic Acid
Grade HPLC