Learn More
2-Azidoethyl 2-Acetamido-2-deoxy-Beta-D-glucopyranoside, TRC
Product Code. 30329670
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329670 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329670 Supplier Toronto Research Chemicals Supplier No. TRCA8489955MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Azidoethyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside
CAS 142072-12-6
Molecular Formula C10H18N4O6
Quantity 5 mg
InChI Formula InChI=1S/C10H18N4O6/c1-5(16)13-7-9(18)8(17)6(4-15)20-10(7)19-3-2-12-14-11/h6-10,15,17-18H,2-4H2,1H3,(H,13,16)/t6-,7-,8-,9-,10-/m1/s1
SMILES CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OCCN=[N+]=[N-]
IUPAC Name N-[(2R,3R,4R,5S,6R)-2-(2-azidoethoxy)-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Molecular Weight (g/mol) 290.28
Formula Weight 290.1226