Learn More
2-Bromo-2’-fluoroacetophenone, TRC
Product Code. 30338233
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338233 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338233 Supplier Toronto Research Chemicals Supplier No. TRCB6843551G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Bromo-2'-fluoroacetophenone
CAS 655-15-2
Molecular Formula C8 H6 Br F O
Quantity 1 g
Synonym Vonoprazan Impurity 96
InChI Formula InChI=1S/C8H6BrFO/c9-5-8(11)6-3-1-2-4-7(6)10/h1-4H,5H2
SMILES Fc1ccccc1C(=O)CBr
IUPAC Name 2-bromo-1-(2-fluorophenyl)ethanone
Molecular Weight (g/mol) 217.04
Formula Weight 215.9586