Learn More
2-Bromobenzyl Bromide, TRC
Product Code. 30339998
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339998 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339998 Supplier Toronto Research Chemicals Supplier No. TRCB67997525G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Bromobenzyl Bromide
CAS 3433-80-5
Molecular Formula C7H6Br2
Quantity 25 g
Synonym α,o-Dibromotoluene, o-Bromobenzyl Bromide, o-(Bromomethyl)bromobenzene, 2-(Bromomethyl)bromobenzene, 1-Bromomethyl-2-bromobenzene, 1-Bromo-2-(bromomethyl)benzene
InChI Formula InChI=1S/C7H6Br2/c8-5-6-3-1-2-4-7(6)9/h1-4H,5H2
SMILES BrCc1ccccc1Br
IUPAC Name 1-bromo-2-(bromomethyl)benzene
Molecular Weight (g/mol) 249.93
Formula Weight 247.8836