Learn More
2-Chloro-10H-phenothiazine, 98%, BLD Pharm™
Product Code. 31332716
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31332716 5 g
31328647 10 g
31329769 25 g
31318755 100 g
31328782 500 g
31327045 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31332716 Supplier BLD Pharm Supplier No. BD143875g
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 92-39-7
Molecular Formula C12H8ClNS
MDL Number MFCD00005016
Quantity 5 g
SMILES ClC(C=C1N2)=CC=C1SC3=C2C=CC=C3
Molecular Weight (g/mol) 233.72
Percent Purity 98%
Chemical Name or Material 2-Chloro-10H-phenothiazine