Learn More
2-Chloro-4-amino-6,7-dimethoxyquinazoline, 98%, BLD Pharm™
Product Code. 30942575
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30942575 100 g 100g
30933760 500 g 500g
30930680 5 g 5g
30936109 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30942575 Supplier BLD Pharm Supplier No. BD11762100g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 23680-84-4
Molecular Formula C10H10ClN3O2
MDL Number MFCD00051734
Quantity 100 g
SMILES NC1=C2C=C(OC)C(OC)=CC2=NC(Cl)=N1
Molecular Weight (g/mol) 239.66
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-amino-6,7-dimethoxyquinazoline