Learn More
2-Chloro Phenothiazine, TRC
Product Code. 30350141
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30350141 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350141 Supplier Toronto Research Chemicals Supplier No. TRCC37497025G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chloro-10H-phenothiazine
CAS 92-39-7
Molecular Formula C12 H8 Cl N S
Quantity 25 g
Synonym 2-Chloro-10H-phenothiazine, Chlorpromazine Hydrochloride Imp. E (EP)
InChI Formula InChI=1S/C12H8ClNS/c13-8-5-6-12-10(7-8)14-9-3-1-2-4-11(9)15-12/h1-7,14H
SMILES Clc1ccc2Sc3ccccc3Nc2c1
IUPAC Name 2-chloro-10H-phenothiazine
Molecular Weight (g/mol) 233.72
Formula Weight 233.0066