Learn More
2-(Cyclohexanecarbonyl)-2,3,6,7-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4(11bH)-one, 98%, BLD Pharm™
Product Code. 31005740
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31005740 5 g 5g
30992403 25 g 25g
30989107 100 g 100g
31003201 10 g 10g
30988856 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31005740 Supplier BLD Pharm Supplier No. BD1490085g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 55268-74-1
Molecular Formula C19H24N2O2
MDL Number MFCD00058531
Quantity 5 g
SMILES O=C1CN(C(C2CCCCC2)=O)CC3N1CCC4=C3C=CC=C4
Molecular Weight (g/mol) 312.41
Percent Purity 98%
Chemical Name or Material 2-(Cyclohexanecarbonyl)-2,3,6,7-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4(11bH)-one