Learn More
2-Ethenyl-2-methyl-5-prop-1-en-2-yloxolane, TRC
Product Code. 30344778
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30344778 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344778 Supplier Toronto Research Chemicals Supplier No. TRCB125313250MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Ethenyl-2-methyl-5-prop-1-en-2-yloxolane
CAS 13679-86-2
Molecular Formula C10H16O
Quantity 250 mg
InChI Formula InChI=1S/C10H16O/c1-5-10(4)7-6-9(11-10)8(2)3/h5,9H,1-2,6-7H2,3-4H3
SMILES CC(=C)C1CCC(C)(O1)C=C
IUPAC Name 2-ethenyl-2-methyl-5-prop-1-en-2-yloxolane
Molecular Weight (g/mol) 152.23
Formula Weight 152.1201