missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester, 96%

Product Code. 15429735
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429735 1 g 1g
15439735 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429735 Supplier Thermo Scientific Alfa Aesar Supplier No. H52312.03
 Restricted Item
Expédition estimée: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1073338-92-7 | C13H19BO5 | 266.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073338-92-7
Molecular Formula C13H19BO5
Molecular Weight (g/mol) 266.10
MDL Number MFCD11855980
InChI Key UMMKKZJPSPIDGN-UHFFFAOYSA-N
PubChem CID 46739337
IUPAC Name ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylate
SMILES CCOC(=O)C1=CC=C(O1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 1 g
Synonym ethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan-2-carboxylate, 5-ethoxycarbonyl furan-2-boronic acid, pinacol ester, 5-ethoxycarbonyl furan-2-boronic acid pinacol ester, 2-ethoxycarbonyl furan-5-boronic acid pinacol ester, ethyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl furan-2-carboxylate, ethyl5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan-2-carboxylate, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-furan-2-carboxylic acid ethyl ester
Formula Weight 266.1
Percent Purity 96%
Chemical Name or Material 2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.