Learn More
2-Fluoro-5-nitrobenzaldehyde, 98%, BLD Pharm™
Product Code. 30948222
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948222 500 g 500g
30932749 25 g 25g
30947810 10 g 10g
30938212 100 g 100g
30930121 5 g 5g
30947266 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30948222 Supplier BLD Pharm Supplier No. BD10307500g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 27996-87-8
Molecular Formula C7H4FNO3
MDL Number MFCD00042298
Quantity 500 g
SMILES C1=C(C(=CC=C1[N+](=O)[O-])F)C=O
Molecular Weight (g/mol) 169.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-nitrobenzaldehyde