Learn More
Gilac™ 2-in-1 Patient Drinking Cup with Nose Cut-out, Green
Product Code. 17918921
Change view
Click to view available options
Capacity (Metric):
0.25 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17918921 0.25 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17918921 Supplier Gilac™ Supplier No. G128508
  • ONLINE EXCLUSIVE PRICE
    3.02€
    1.99€ / 1 pièces
    Économisez 1.03€
    34% de réduction
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Green 2-in-1 Patient Drinking Cup with Nose Cut-out allows drinking without spilling content, either with the straw or thanks to the cut-out nosed shape.
  • Drinking cup with graduations (in mL).
  • Integrated lid and drinking straw.
  • Allows drinking without spilling content.
Specifications
Material Polypropylene
Capacity (Metric) 0.25 L
Capacity (English) 8.5 oz.
Product Type Tumbler Cup