Learn More
2-Methoxyphenothiazine, 96%, BLD Pharm™
Specifications
Specifications
| CAS | 1771-18-2 |
| Molecular Formula | C13H11NOS |
| MDL Number | MFCD00037838 |
| Quantity | 5 g |
| SMILES | COC(C=C1N2)=CC=C1SC3=C2C=CC=C3 |
| Molecular Weight (g/mol) | 229.30 |
| Percent Purity | 96% |
| Chemical Name or Material | 2-Methoxyphenothiazine |