Learn More
2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic Acid, TRC
Product Code. 30466033
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30466033 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466033 Supplier Toronto Research Chemicals Supplier No. TRCM3271601G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic Acid
CAS 739364-95-5
Molecular Formula C8H7N3O2
Quantity 1 g
InChI Formula InChI=1S/C8H7N3O2/c1-5-2-7-9-3-6(8(12)13)4-11(7)10-5/h2-4H,1H3,(H,12,13)
SMILES O=C(C1=CN2N=C(C)C=C2N=C1)O
IUPAC Name 2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Molecular Weight (g/mol) 177.16
Formula Weight 177.05