missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)ethylamine, Thermo Scientific™

Product Code. 10486652
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10486652 10 g Amber Glass Bottle 10g
10511872 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10486652 Supplier Thermo Scientific Maybridge Supplier No. CD06838EA
 Restricted Item
Expédition estimée: 03-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 18542-42-2
Molecular Formula C3H9NS
Molecular Weight (g/mol) 91.172
MDL Number MFCD00014825
InChI Key CYWGSFFHHMQKET-UHFFFAOYSA-N
Synonym 2-methylthio ethylamine, 2-methylthio ethanamine, 2-aminoethyl methyl sulfide, 2-methylsulfanyl ethanamine, ethanamine, 2-methylthio, 2-methylsulfanyl ethan-1-amine, 2-methylthio ethyl amine, ethylamine, 2-methylthio, s-methylcysteamine
PubChem CID 87697
IUPAC Name 2-methylsulfanylethanamine
SMILES CSCCN

Specifications

CAS Min % 80.0
CAS Max % 100.0
Density 0.974g/mL
Assay Percent Range 95%+
Packaging Amber Glass Bottle
Linear Formula CH3S(CH2)2NH2
Quantity 10 g
Formula Weight 91.18
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-(Methylthio)ethylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.