Learn More
2-Nitro-1,4-bis(trifluoromethyl)benzene, 97%, BLD Pharm™
Product Code. 30962301
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30962301 5 g 5g
30955597 25 g 25g
30959163 10 g 10g
30961442 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30962301 Supplier BLD Pharm Supplier No. BD108045g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 320-88-7
Molecular Formula C8H3F6NO2
MDL Number MFCD00236674
Quantity 5 g
SMILES [O-][N+](=O)C1=C(C=CC(=C1)C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 259.11
Percent Purity 97%
Chemical Name or Material 2-Nitro-1,4-bis(trifluoromethyl)benzene