Learn More
(2-Oxopyrrolidin-1-yl)acetic Acid, LoGiCal
Product Code. 30459870
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459870 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459870 Supplier LoGiCal Supplier No. LGCFOR0083.01
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2-Oxopyrrolidin-1-yl)acetic Acid
CAS 53934-76-2
Analyte or Component Names (2-Oxopyrrolidin-1-yl)acetic Acid
Shipping Condition Room Temperature
Molecular Formula C6 H9 N O3
Certifications/Compliance ISO 9001
Synonym (2-Oxopyrrolidin-1-yl)acetic Acid, Piracetam Imp. D (EP), 2-Oxo-1-pyrrolidineacetic acid, (2-Oxopyrrolidin-1-yl)acetic acid, 2-Oxo-1-pyrrolidineacetic acid, 2-Pyrrolidinon-1-ylacetic acid, 2-Pyrrolidinoneacetic acid, Piracetam Impurity D
InChI Formula InChI=1S/C6H9NO3/c8-5-2-1-3-7(5)4-6(9)10/h1-4H2,(H,9,10)
Solution Type Neat
SMILES OC(=O)CN1CCCC1=O
IUPAC Name 2-(2-oxopyrrolidin-1-yl)acetic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 143.14
Quantity 10 mg
Formula Weight 143.0582 g/mol